Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gstp1
Entrez Gene ID
100689311
Gene Name
glutathione S-transferase, pi 1
Gene Aliases
I79_018157
Chromosome Location
Chr.Un|NW_003614797: 143919 - 146124 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246744.1 NP_001233673.1 3 - 4 229 76
GenBank mRNA JI886703.1 - 4 - 5 328 76
JP038870.1 - 4 - 5 328 76
L40381.1 - 3 - 4 229 76

View Full Product Details