Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss41
Entrez Gene ID
100772113
Gene Name
protease, serine, 41
Gene Aliases
I79_022428
Chromosome Location
Chr.Un|NW_003615934: 37402 - 41844 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513456.1 XP_003513504.1 2 - 3 130 78

View Full Product Details