Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd61
Entrez Gene ID
100756204
Gene Name
ankyrin repeat domain 61
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613618: 71963 - 77496 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496275.2 XP_003496323.1 1 - 2 821 71

View Full Product Details