Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fosl2
Entrez Gene ID
100755919
Gene Name
FOS-like antigen 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613628: 2292733 - 2313826 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496572.2 XP_003496620.1 3 - 4 1225 92

View Full Product Details