Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Id2
Entrez Gene ID
100769110
Gene Name
inhibitor of DNA binding 2, dominant negative helix-loop-helix protein
Gene Aliases
I79_011789
Chromosome Location
Chr.Un|NW_003614082: 495246 - 497519 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504693.2 XP_003504741.1 1 - 2 409 63
GenBank mRNA JP061365.1 - 1 - 2 419 63

View Full Product Details