Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100751713
Entrez Gene ID
100751713
Gene Name
chromosome unknown open reading frame, human C15orf43
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613677: 115652 - 138570 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498025.1 XP_003498073.1 2 - 3 145 100

View Full Product Details