Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spatc1
Entrez Gene ID
100752755
Gene Name
spermatogenesis and centriole associated 1
Gene Aliases
I79_015372, Speriolin
Chromosome Location
Chr.Un|NW_003614411: 469608 - 493185 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507627.1 XP_003507675.1 4 - 5 1318 78

View Full Product Details