Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clec2l
Entrez Gene ID
100770823
Gene Name
C-type lectin domain family 2, member L
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613951: 269218 - 304075 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503229.2 XP_003503277.2 2 - 3 113 68

View Full Product Details