Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmgb3
Entrez Gene ID
100772377
Gene Name
high mobility group box 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614816: 427315 - 429268 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510038.2 XP_003510086.1 1 - 2 160 90

View Full Product Details