Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnrip1
Entrez Gene ID
100773416
Gene Name
cannabinoid receptor interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613893: 633410 - 654516 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502379.2 XP_003502427.1 1 - 2 512 64
GenBank mRNA JI888983.1 - 1 - 2 97 64
JP053821.1 - 1 - 2 280 64

View Full Product Details