Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100754798
Entrez Gene ID
100754798
Gene Name
chromosome unknown open reading frame, human C19orf26
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613904: 916075 - 922958 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643250.1 XP_007641440.1 4 - 5 610 109

View Full Product Details