Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss38
Entrez Gene ID
100764762
Gene Name
protease, serine, 38
Gene Aliases
I79_010138
Chromosome Location
Chr.Un|NW_003613960: 598961 - 610760 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503353.1 XP_003503401.1 4 - 5 715 105

View Full Product Details