Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr52
Entrez Gene ID
100766957
Gene Name
WD repeat domain 52
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613667: 1566191 - 1644136 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497857.1 XP_003497905.1 31 - 32 5032 85

View Full Product Details