Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dbndd2
Entrez Gene ID
100765248
Gene Name
dysbindin (dystrobrevin binding protein 1) domain containing 2
Gene Aliases
I79_011678
Chromosome Location
Chr.Un|NW_003614077: 884870 - 914929 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645109.1 XP_007643299.1 2 - 3 699 72
XM_007645110.1 XP_007643300.1 2 - 3 677 72
XM_007645111.1 XP_007643301.1 2 - 3 651 72
XM_007645112.1 XP_007643302.1 1 - 2 153 72
XM_007645113.1 XP_007643303.1 2 - 3 297 72
XM_007645114.1 XP_007643304.1 2 - 3 347 72
GenBank mRNA JP063969.1 - 1 - 2 134 72

View Full Product Details