Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
G6pc2
Entrez Gene ID
100762184
Gene Name
glucose-6-phosphatase, catalytic, 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613606: 430649 - 437404 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495933.1 XP_003495981.1 4 - 5 554 94

View Full Product Details