Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdh19
Entrez Gene ID
100774560
Gene Name
protocadherin 19
Gene Aliases
I79_005735
Chromosome Location
Chr.Un|NW_003613747: 1200248 - 1309444 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640375.1 XP_007638565.1 6 - 7 2758 80

View Full Product Details