Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim40
Entrez Gene ID
100753736
Gene Name
tripartite motif containing 40
Gene Aliases
I79_015094
Chromosome Location
Chr.Un|NW_003614381: 448540 - 455269 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507430.1 XP_003507478.1 4 - 5 676 86

View Full Product Details