Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rerg
Entrez Gene ID
100756816
Gene Name
RAS-like, estrogen-regulated, growth inhibitor
Gene Aliases
I79_005247
Chromosome Location
Chr.Un|NW_003613723: 2012092 - 2108555 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007639689.1 XP_007637879.1 1 - 2 58 79

View Full Product Details