Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4d
Entrez Gene ID
100775089
Gene Name
C-type lectin domain family 4, member D
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614841: 355281 - 378030 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510185.2 XP_003510233.1 2 - 3 127 63

View Full Product Details