Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Odf3l1
Entrez Gene ID
100760080
Gene Name
outer dense fiber of sperm tails 3-like 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613669: 2511439 - 2522093 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497916.2 XP_003497964.1 4 - 5 408 88

View Full Product Details