Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekhg7
Entrez Gene ID
103162196
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614907: 370923 - 384824 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651202.1 XP_007649392.1 2 - 3 171 106

View Full Product Details