Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkx6-3
Entrez Gene ID
100751652
Gene Name
NK6 homeobox 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613958: 1120835 - 1124611 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503318.1 XP_003503366.1 1 - 2 381 92

View Full Product Details