Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scrt2
Entrez Gene ID
100761306
Gene Name
scratch family zinc finger 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615132: 200792 - 213089 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652056.1 XP_007650246.1 3 - 4 640 94

View Full Product Details