Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Apoc2
Entrez Gene ID
100774395
Gene Name
apolipoprotein C-II
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614150: 289771 - 290589 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505455.1 XP_003505503.1 2 - 3 207 59
XM_007645950.1 XP_007644140.1 2 - 3 216 59

View Full Product Details