Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcp10l2
Entrez Gene ID
100772850
Gene Name
t-complex 10-like 2
Gene Aliases
I79_011049
Chromosome Location
Chr.Un|NW_003614028: 1200628 - 1220057 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504054.2 XP_003504102.1 5 - 6 747 81

View Full Product Details