Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam83c
Entrez Gene ID
100761734
Gene Name
family with sequence similarity 83, member C
Gene Aliases
I79_007511
Chromosome Location
Chr.Un|NW_003613833: 1530057 - 1537401 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641836.1 XP_007640026.1 1 - 2 847 71

View Full Product Details