Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gas2l2
Entrez Gene ID
100774805
Gene Name
growth arrest-specific 2 like 2
Gene Aliases
I79_019780
Chromosome Location
Chr.Un|NW_003615071: 141335 - 148754 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511301.1 XP_003511349.1 4 - 5 832 88

View Full Product Details