Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cidea
Entrez Gene ID
100763404
Gene Name
cell death-inducing DFFA-like effector a
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614106: 452494 - 483250 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645435.1 XP_007643625.1 3 - 4 408 77

View Full Product Details