Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Leprel4
Entrez Gene ID
100772453
Gene Name
leprecan-like 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614078: 90361 - 96657 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504621.2 XP_003504669.1 1 - 2 149 62

View Full Product Details