Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mogat3
Entrez Gene ID
100769277
Gene Name
monoacylglycerol O-acyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613822: 1827974 - 1831897 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641693.1 XP_007639883.1 3 - 4 253 99

View Full Product Details