Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcl2l10
Entrez Gene ID
100752860
Gene Name
BCL2-like 10 (apoptosis facilitator)
Gene Aliases
I79_000127
Chromosome Location
Chr.Un|NW_003613582: 1217687 - 1221006 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495054.1 XP_003495102.1 1 - 2 426 110

View Full Product Details