Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pawr
Entrez Gene ID
100754431
Gene Name
PRKC, apoptosis, WT1, regulator
Gene Aliases
I79_016311
Chromosome Location
Chr.Un|NW_003614534: 340548 - 374430 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508386.2 XP_003508434.1 1 - 2 129 78

View Full Product Details