Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndufb4
Entrez Gene ID
100762129
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613831: 550298 - 557011 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501085.2 XP_003501133.1 2 - 3 428 83
GenBank mRNA JI885867.1 - 2 - 3 360 83
JP064278.1 - 2 - 3 360 83

View Full Product Details