Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arx
Entrez Gene ID
100759833
Gene Name
aristaless related homeobox
Gene Aliases
I79_016079
Chromosome Location
Chr.Un|NW_003614500: 339140 - 355079 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508210.2 XP_003508258.1 5 - 6 1153 103

View Full Product Details