Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dact2
Entrez Gene ID
100774769
Gene Name
dishevelled-binding antagonist of beta-catenin 2
Gene Aliases
I79_011041
Chromosome Location
Chr.Un|NW_003614028: 614309 - 671592 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644579.1 XP_007642769.1 2 - 3 213 109

View Full Product Details