Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMH4
Entrez Gene ID
102191431
Gene Name
armadillo like helical domain containing 4
Gene Aliases
C10H14orf37
Chromosome Location
Chr.10: 32752736 - 32894604 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054095.1 XP_017909584.1 2 - 3 1596 79
XM_018054096.1 XP_017909585.1 2 - 3 1597 79
GenBank mRNA KA315308.1 - 6 - 7 1628 79

View Full Product Details