Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC23
Entrez Gene ID
102187264
Gene Name
zinc finger DHHC-type containing 23
Gene Aliases
-
Chromosome Location
Chr.1: 58387275 - 58399787 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039430.1 XP_017894919.1 2 - 3 436 78
XM_018039495.1 XP_017894984.1 2 - 3 437 78
GenBank mRNA JO434432.2 - 2 - 3 226 78
JO445416.2 - 2 - 3 226 78
JO591579.2 - 2 - 3 353 78
JO591580.2 - 2 - 3 353 78
JT705700.1 - 3 - 4 304 78
JT717393.1 - 2 - 3 318 78
KA353112.1 - 2 - 3 259 78

View Full Product Details