Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PJVK
Entrez Gene ID
102174249
Gene Name
pejvakin
Gene Aliases
DFNB59
Chromosome Location
Chr.2: 117882558 - 117889068 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038786.1 XP_017894275.1 3 - 4 542 69

View Full Product Details