Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAG4
Entrez Gene ID
102179734
Gene Name
BCL2 associated athanogene 4
Gene Aliases
-
Chromosome Location
Chr.27: 12289416 - 12321466 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041795.1 XP_017897284.1 4 - 5 1005 80
GenBank mRNA KA344631.1 - 4 - 5 735 80

View Full Product Details