Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NLGN3
Entrez Gene ID
102168988
Gene Name
neuroligin 3
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 10597644 - 10613688 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043873.1 XP_017899362.1 2 - 3 920 72

View Full Product Details