Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ZBTB45
Entrez Gene ID
106503173
Gene Name
zinc finger and BTB domain containing 45
Gene Aliases
-
Chromosome Location
Chr.18: 67168954 - 67173992 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063536.1 XP_017919025.1 1 - 2 167 67

View Full Product Details