Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF19
Entrez Gene ID
102181149
Gene Name
zinc finger protein 19
Gene Aliases
-
Chromosome Location
Chr.18: 40783916 - 40794374 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692223.3 XP_005692280.2 2 - 3 97 61

View Full Product Details