Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H6PD
Entrez Gene ID
102174863
Gene Name
hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.16: 42698677 - 42738707 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060387.1 XP_017915876.1 3 - 4 973 79
XM_018060388.1 XP_017915877.1 3 - 4 1165 79
GenBank mRNA JO425030.1 - 2 - 3 3161 79
JO425043.1 - 2 - 3 3161 79
JO425188.1 - 2 - 3 3161 79

View Full Product Details