Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLEC2L
Entrez Gene ID
102177690
Gene Name
C-type lectin domain family 2 member L
Gene Aliases
-
Chromosome Location
Chr.4: 17394053 - 17410613 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046877.1 XP_017902366.1 2 - 3 264 68
GenBank mRNA JO644397.2 - 2 - 3 234 68
JO644398.2 - 2 - 3 234 68

View Full Product Details