Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR88
Entrez Gene ID
102189788
Gene Name
WD repeat domain 88
Gene Aliases
-
Chromosome Location
Chr.18: 44603694 - 44656107 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692488.2 XP_005692545.2 5 - 6 723 58

View Full Product Details