Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLIPR1L1
Entrez Gene ID
102177408
Gene Name
GLIPR1 like 1
Gene Aliases
-
Chromosome Location
Chr.5: 4643056 - 4679156 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679728.2 XP_005679785.2 4 - 5 718 90

View Full Product Details