Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102179170
Entrez Gene ID
102179170
Gene Name
putative tetratricopeptide repeat protein 41
Gene Aliases
-
Chromosome Location
Chr.5: 66205797 - 66287937 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680542.3 XP_005680599.2 8 - 9 3036 67

View Full Product Details