Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TERB2
Entrez Gene ID
102168835
Gene Name
telomere repeat binding bouquet formation protein 2
Gene Aliases
-
Chromosome Location
Chr.10: 48724 - 85520 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686226.2 XP_005686283.2 3 - 4 309 90

View Full Product Details