Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARRDC5
Entrez Gene ID
102184140
Gene Name
arrestin domain containing 5
Gene Aliases
-
Chromosome Location
Chr.7: 91272965 - 91292090 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682519.3 XP_005682576.2 3 - 4 2500 59

View Full Product Details