Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THEMIS
Entrez Gene ID
102185656
Gene Name
thymocyte selection associated
Gene Aliases
-
Chromosome Location
Chr.9: 53018390 - 53225408 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684724.2 XP_005684781.2 2 - 3 361 92

View Full Product Details